Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a protein which is necessary for normal skeletal development and brain function. Mutations in this gene are associated with two types of recessive osteochondrodysplasia, Dyggve-Melchior-Clausen (DMC) dysplasia and Smith-McCort (SMC) dysplasia, which involve both skeletal defects and mental retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dyggve-Melchior-Clausen syndrome protein; Dymeclin
Gene Aliases: 1810041M12Rik; 4933427L07Rik; C030019K18Rik; DMC; DYM; RGD1309111; SMC
UniProt ID: (Human) Q7RTS9, (Mouse) Q8CHY3, (Rat) B4F766
Entrez Gene ID: (Human) 54808, (Mouse) 69190, (Rat) 291433
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support