Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RSNQMNKKK TSGNQDEILV IRKGWLTINN IGIMKGGSKE YWFVLTAENL SWYKDDEEKE KKYMLSVDNL KLRDVEKGFM SSKHIFALFN TEQRNVYKDY RQLELACETQ EEVDSWKASF LRAGVYPERV GDKEKASETE ENGSDSFMHS MDPQLERQVE TIRNLVDSYM AIVNKTVRDL MPKTIMHLMI NNTKEFIFSE LLANLYSCGD QNTLMEESAE QAQRRDEMLR MYHALKEALS IIGDINTTTV STPMPPPVDD SWLQVQSVPA GRRSPTSSPT PQRRAPAVPP ARPGSRGPAP GPPPAGSALG GAPPVPSRPG ASPDPFGPPP QVPSRPNRAP PGVPRITISD P (502-851 aa encoded by BC063850)
Dynamin is a GTPase required for endocytosis of clathrin coated vesicles from the membrane. Dynamin's involvement in endocytosis was first discovered during the characterization of the temperature sensitive parylytic shibire mutation from Drosophila melanogaster. Three closely related isoforms have been identified in mammals. Dynamin1 is expressed exclusively in neurons, dynamin2 is ubiquitously expressed and dynamin3 is expressed primarily in the testes. Each of these isoforms of dynamin also has several splice variants. This suggests that each isoform/splice variant may have separate intracellular targets or function in the budding of distinct vesicle types. Common domain features of the dynamin proteins include an amino acid terminal GTPase domain, a central pleckstrin homology domain and a highly basic proline rich carboxyl terminus with several Src homology 3 (SH3) domains. These SH3 domains target the dynamin to the clathrin coated pit. Dynamin is known to interact with at least two proteins, AP2 adaptor protein complex and amphiphysin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-dynamin; D100; Dyn1; Dynamin; Dynamin BREDNM19; Dynamin I; Dynamin, brain; Dynamin-1; Dynamin1; EC 3.6.5.5; Neuronal dynamin; OTTHUMP00000215500
Gene Aliases: AI838169; DNM; DNM1; EIEE31; Ftfl; Kiaa4093; mKIAA4093
UniProt ID: (Human) Q05193, (Mouse) P39053, (Rat) P21575
Entrez Gene ID: (Human) 1759, (Mouse) 13429, (Rat) 140694
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support