Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: VGEKSWVQF NSNSQLMYGL PDSSHVGKHE YFMHATDKGG LSAVDAFEIH VHRRPQGDRA PARFKAKFVG DPALVLNDIH KKIALVKKLA FAFGDRNCST ITLQNITRGS IVVEWTNNTL PLEPCPKEQI AGLSRRIAED DGKPRPAFSN ALEPDFKATS ITVTGSGSCR HLQFIPVVPP RRVPSEAPPT EVPDRDPEKS SEDDVYLHTV IPAVVVAAIL LIAGIIAMIC YRKKRKGKLT LEDQATFIKK GVPIIFADEL DDSKPPPSSS MPLILQEEKA PLPPPEYPNQ SVPETTPLNQ DTMGEYTPLR DEDPNAPPYQ PPPPFTAPME GKGSRPKNMT PYRSPPPYVP P (546-895 aa encoded by BC012740)
Dystroglycan is a laminin binding component of the dystrophin-glycoprotein complex which provides a linkage between the subsarcolemmal cytoskeleton and the extracellular matrix. Dystroglycan 1 is a candidate gene for the site of the mutation in autosomal recessive muscular dystrophies. The dramatic reduction of dystroglycan 1 in Duchenne muscular dystrophy leads to a loss of linkage between the sarcolemma and extracellular matrix, rendering muscle fibers more susceptible to necrosis. Dystroglycan also functions as dual receptor for agrin and laminin-2 in the Schwann cell membrane. The muscle and nonmuscle isoforms of dystroglycan differ by carbohydrate moieties but not protein sequence. Alternative splicing results in multiple transcript variants all encoding the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-dystroglycan; beta-dystroglycan; Dystroglycan; Dystroglycan 1; dystroglycan 1 (dystrophin-associated glycoprotein 1); dystrophin associated glycoprotein 1; Dystrophin-associated glycoprotein 1
Gene Aliases: 156DAG; A3a; AGRNR; D9Wsu13e; DAG; DAG1; DG; Dp427; Dp71; MDDGA9; MDDGC7; MDDGC9
UniProt ID: (Human) Q14118, (Mouse) Q62165
Entrez Gene ID: (Human) 1605, (Pig) 100620392, (Mouse) 13138, (Rat) 114489
Molecular Function:
cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support