Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KCDPGFSGLK CEQIVNCTAL ESPEHGSLVC SHPLGNFSYN SSCSISCDRG YLPSSMETMQ CMSSGEWSAP IPACNVVECD AVTNPANGFV ECFQNPGSFP WNTTCTFDCE EGFELMGAQS LQCTSSGNWD NEKPTCKAVT CRAVRQPQNG SVRCSHSPAG EFTFKSSCNF TCEEGFMLQG PAQVECTTQG QWTQQIPVCE AFQCTALS
Leukocytes adhere to the blood vessel endothelium during extravasation in postcapillary venules of lymph nodes. In addition, leukocyte adhesion occurs in the capillaries and small venules at any site in the body after onset of inflammation. This response is immediate and involves a cascade of adhesion receptors. At the endothelial surface members of the selectin and immunoglobulin superfamilies participate in this cascade. Selectins are C-type cell surface lectins that play a role in leukocyte adhesion to the blood vessel wall endothelium. E-selectin (CD62E) is an endothelial cell specific selectin that is expressed only after activation with proinflammatory cytokines. In vitro experiments have shown that IL-1, TNFalpha and bacterial wall components like lipopolysaccharides induce the transcription of E-selectin in a NFkB dependent signaling cascade. E-selectin has been associated with blood vessel endothelium in diverse inflammatory situations.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD62 antigen-like family member E; CD62E; e selectin; E-selectin; ELAM-1; endothelial adhesion molecule 1; Endothelial leukocyte adhesion molecule 1; LECAM2; leukocyte endothelial cell adhesion molecule 2; Leukocyte-endothelial cell adhesion molecule 2; RP1-117P20.2
Gene Aliases: CD62E; ELAM; ELAM1; ESEL; LECAM2; SELE
UniProt ID: (Human) P16581
Entrez Gene ID: (Human) 6401
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support