OriGene
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This antibody also recognizes DsRed-Monomer protein.
E2-Crimson fluorescent protein.
Protein Aliases: E2-Crimson