Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: PPEDLLQSPS AVSTPPPLPK PALAQSQEAS RPNSPQLTPT AVPGSAEVQG MAGPAAEITV SGGPGTDSKD SGELSSLPLG PTTLDTRPLQ
E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-4 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides Differential phosphorylation contributes to the microheterogeniety of E2F-4 (total aa 411-416, varies due to a trinucleotide repeat).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E2F transcription factor 4, p107/p130-binding; E2F-4; p107/p130-binding protein; Transcription factor E2F4
Gene Aliases: E2F-4; E2F4
UniProt ID: (Human) Q15328
Entrez Gene ID: (Human) 1874
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support