Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAQLAAICKM QLEEQSSESR QKVKVQLARS GPCKPVAPLD PPVNAEMELT APSLIQPLGM VPLIPSPLSS AVPLILPQAP SGPSYAIYLQ PTQAHQSVTP PQGLSPTVCT THSSKATGSK DSTDATTEKA ANDTSKASAS TRPGSLLPAP ERQGAKSRTR EPAGERGSKR ASMLEDSGSK KKFKEDLKGL ENVSATLFPS GYLIPLTQCS SLGAESILSG KENSSALSPN HRIYSSPIAG VIPVTSSELT AVNFPSFHVT PLKLMVSPTS VAAVPVGNSP ALASSHPVPI QNPSSAIVNF TLQHLGLISP NVQLSASPGS GIVPVSPRIE SVNVAPENAG TQQGRATNYD SPVPGQSQPN GQSVAVTGAQ QPVPVTPKGS QLVAESFFRT PGGPTKPTSS SCMDFEGANK TSLGTLFVPQ RKLEVSTEDV H
E2F transcription factors, such as E2F8, are essential for orchestrating expression of genes required for cell cycle progression and proliferation (Christensen et al., 2005).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E2F 8; E2F family member 8; E2F-8; FLJ23311; Transcription factor E2F8
Gene Aliases: E2F-8; E2F8
UniProt ID: (Human) A0AVK6
Entrez Gene ID: (Human) 79733
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support