Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Excitatory amino acid transporter 2 (EAAT2) is a member of a family of solute transporter proteins. The membrane-bound protein is the principal transporter that clears the excitatory neurotransmitter glutamate from the extracellular space at synapses in the central nervous system. Glutamate clearance is necessary for proper synaptic activation and to prevent neuronal damage from excessive activation of glutamate receptors. Mutations in and decreased expression of this protein are associated with amyotrophic lateral sclerosis (ALS.) Alternatively spliced transcript variants of this gene have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EAAT2 variant mEAAT2/5UT5; Excitatory amino acid transporter 2; excitotoxic amino acid transporter 2; glial high affinity glutamate transporter; GLT-1; GLT1a; GLT1b; GLT1c; GLUT-R; glutamate transporter 1c; Glutamate/aspartate transporter II; high affinity glucose transporter; OTTHUMP00000165209; OTTHUMP00000165210; Sodium-dependent glutamate/aspartate transporter 2; solute carrier family 1 (glial high affinity glutamate transporter), member 2; Solute carrier family 1 member 2; solute carrier family 1, member 2
Gene Aliases: 1700091C19Rik; 2900019G14Rik; AI159670; EAAT2; Glt; GLT-1; GLT1; HBGT; MGLT1; SLC1A2
UniProt ID: (Human) P43004, (Mouse) P43006, (Rat) P31596
Entrez Gene ID: (Human) 6506, (Mouse) 20511, (Rat) 29482
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support