Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MNGTANPLLD REEHCLRLGE SFEKRPRASF HTIRYDFKPA SIDTSCEGEL QVGKGDEVTI TLPHIPGSTP PMTVFKGNKR PYQKDCVLII NHDTGEFVLE KLSSSIQVKK TRAEGSSKIQ ARMEQQPTRP PQTSQPPPPP PPMPFRAPTK PPVGPKTSPL KDNPSPEPQL DDIKRELRAE VDIIEQMSSS SGSSSSDSES SSGSDDDSSS SGGEDNGPAS PPQPSHQQPY NSRPAVANGT SRPQGSNQLM NALRNDLQLS ESGSDSDD
Acts as a transcriptional transactivator of ELL and ELL2 elongation activities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ELL (eleven nineteen lysine-rich leukemia gene)-associated factor 1; ELL-associated factor 1
Gene Aliases: EAF1
UniProt ID: (Human) Q96JC9
Entrez Gene ID: (Human) 85403
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support