Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TAKHLLLYQA LGWQPPHFAH LPLLLNRDGS KLSKRQGDVF LEHFAADGFL
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 100%; Yeast: 91%; Zebrafish: 93%
Glutamyl-tRNA synthetase (GluRS or EARS2) a class I aminoacyl-tRNA synthetase (aaRS), is primarily responsible for the glutamylation of tRNAGlu. It is part of the "minimal set" of seventeen aaRSs found in every living organism and its presence is essential for the viability of cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GluRS; glutamate tRNA ligase 2, mitochondrial; glutamate--tRNA ligase; Glutamate--tRNA(Gln) ligase EARS2, mitochondrial; Glutamyl-tRNA synthetase; Mitochondrial glutamyl-tRNA synthetase; mtGluRS; Nondiscriminating glutamyl-tRNA synthetase EARS2, mitochondrial; probable glutamyl-tRNA synthetase, mitochondrial
Gene Aliases: COXPD12; EARS2; gluRS; KIAA1970; MSE1
UniProt ID: (Human) Q5JPH6
Entrez Gene ID: (Human) 124454
Molecular Function:
aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support