Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Glutamyl-tRNA synthetase (GluRS or EARS2) a class I aminoacyl-tRNA synthetase (aaRS), is primarily responsible for the glutamylation of tRNAGlu. It is part of the "minimal set" of seventeen aaRSs found in every living organism and its presence is essential for the viability of cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GluRS; glutamate tRNA ligase 2, mitochondrial; glutamate--tRNA ligase; Glutamate--tRNA(Gln) ligase EARS2, mitochondrial; Glutamyl-tRNA synthetase; glutamyl-tRNA synthetase 2 mitochondrial (putative); Mitochondrial glutamyl-tRNA synthetase; mtGluRS; Nondiscriminating glutamyl-tRNA synthetase EARS2, mitochondrial; probable glutamate--tRNA ligase, mitochondrial; probable glutamyl-tRNA synthetase, mitochondrial
Gene Aliases: 3230401I01Rik; COXPD12; EARS2; gluRS; KIAA1970; mKIAA1970; MSE1; RGD1307904
UniProt ID: (Human) Q5JPH6, (Mouse) Q9CXJ1
Entrez Gene ID: (Human) 124454, (Rat) 361641, (Mouse) 67417
Molecular Function:
aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support