Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat, Chicken and Orangutan
EB1 family proteins are evolutionarily conserved proteins that bind microtubule plus-ends and centrosomes and regulate the dynamics and organization of microtubules. The MAPRE genes encode the EB1 family proteins. The human MAPRE genes MAPRE1, MAPRE2, and MAPRE3 code for EB1, RP1, and EBF3 proteins, respectively. EB1 family members appear to be expressed ubiquitously. The EB1 (MAPRE1) gene, at 20q11. 2, is fused to MLL in an adult patient with pro-B acute lymphoblastic leukemia. EB1 is a microtubule-associated protein that interacts with the colorectal adenomatous polyposis coli tumor-suppressor protein and plays important roles in regulating microtubule dynamics, cell polarity, and chromosome stability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adenomatosis polyposis coli binding protein Eb1; adenomatous polyposis coli-binding protein EB1; APC-binding protein EB1; EB1; End-binding protein 1; MGC117374; MGC129946; Microtubule-associated protein RP/EB family member 1; microtubule-associated protein, RP/EB family, member 1
Gene Aliases: 5530600P05Rik; AI462499; AI504412; AW260097; BIM1p; D2Ertd459e; EB1; MAPRE1
UniProt ID: (Human) Q15691, (Mouse) Q61166
Entrez Gene ID: (Human) 22919, (Mouse) 13589
Molecular Function: non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support