Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: LYGMPHNNQE IILKRAADIA EALYSVPRNH NQIPTLGNNP AHTGMMGVNS FSSQLAVNVS ETSQANDQVG YSRNTSSVSP RGYVPSSTPQ QSNYNTVSTS
Transcriptional activator which recognizes variations of the palindromic sequence 5'-ATTCCCNNGGGAATT-3'.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Early B-cell factor 3; EBF-3; O/E-2; Olf-1/EBF-like 2; Transcription factor COE3
Gene Aliases: COE3; EBF-3; EBF3; O/E-2; OE-2
UniProt ID: (Human) Q9H4W6
Entrez Gene ID: (Human) 253738, (Rat) 361668
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support