Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MPSSPPLAAA SSMSLPAAAP TTSVFSFSPV NMISAVKQRS AFAPVLRPPS SPPQACPRAH GEGLPDQSFE DSDKFHSPAR GLQGLAYS
EBF4 belongs to the COE family and contains 1 IPT/TIG domain. EBF4 is a transcriptional factor which recognizes variations of the palindromic sequence 5'-ATTCCCNNGGGAATT-3'. EBF4 belongs to the conserved Olf/EBF family of helix-loop-helix transcription factors, members of which play important roles in neural development and B-cell maturation (Wang et al., 2002 [PubMed 12139918]).[supplied by OMIM].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Early B-cell factor 4; EBF-4; O/E-4; OE-4; Olf-1/EBF-like 4; RP5-860F19.3; Transcription factor COE4
Gene Aliases: COE4; EBF4; KIAA1442; O/E-4
UniProt ID: (Human) Q9BQW3
Entrez Gene ID: (Human) 57593
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support