Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTPQLLLAL VLWASCPPCS GRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK (1-229 aa encoded by BC015364)
EBI3 (Epstein-Barr virus Induced-3) is a secreted glycoprotein of the hematopoietin receptor family. It plays a critical regulatory role in the induction of Th2-type immune responses and the development of Th2-mediated tissue inflammation in vivo, which may be mediated through the control of iNKT cell function. EBI3 dimerizes with p28 and p35 subunits of IL-12 to form new proteins IL-27 and IL-35, respectively. IL-27 is an early product of activated antigen presenting cell that is produced upon TLR ligation. It negatively regulates Th17 cell differentiation. EBI3 is widely expressed and its expression in dendritic cell is transcriptionally regulated by TLR signaling via MyD88 and NF-kappaB during innate immune responses preceding cytokine driven Th cell development. EBI3 signal inhibits delayed-type hypersensitivity responses by suppressing IL-17 production and inducing IL-10 hyperproduction. EBI3 may play a novel role in controlling tumor metastasis via lung CD8+ T cells, and its deficiency is associated with a diminished production of Th2 cytokines which are known to regulate allergic airway inflammation in asthma. Targeted deletion of EBI3 protects mice from lung metastasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytokine receptor; EBV-induced gene 3 protein; Epstein-Barr virus induced gene 3; Epstein-Barr virus induced gene 3 (similar to Interleukin-27 beta chain precursor); Epstein-Barr virus-induced gene 3 protein; IL-27 subunit beta; IL27 subunit; IL35 subunit; Interleukin-27 subunit beta
Gene Aliases: EBI3; IL-27B; IL27B
UniProt ID: (Human) Q14213
Entrez Gene ID: (Pig) 100522599, (Human) 10148, (Rat) 680609
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support