Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDTPPLSDSE SESDESLVTD RELQDAFSRG LLKPGLNVVL EGPKKAVNDV NGLKQCLAEF KRDLEWVERL DVTLGPVPEI GGSEAPAPQN KDQKAVDPED DFQREMSFYR QAQAAVLAVL PRLHQLKVPT KRPTDYFAEM AKSDLQMQKI RQKLQTKQAA MERSEKAKQL RALRKYGKKV QTEVLQKRQQ EKAHMMNAIK KYQKGFSDKL DFLEGDQKPL AQRKKAGAKG QQMRKGPSAK RRYKNQKFGF GGKKKGSKWN TRESYDDVSS FRAKTAHGRG LKRPGKKGSN KRPGKRTREK MKNRTH; Positive Samples: A-431, LO2, HeLa, HT-1080, K-562, MCF7, Mouse pancreas; Cellular Location: Nucleus, nucleolus
EBNA1BP2 is required for the processing of the 27S pre-rRNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EBNA1-binding protein 2; nuclear FGF3 binding protein; Nucleolar protein p40; nucleolar protein p40; homolog of yeast EBNA1-binding protein; Probable rRNA-processing protein EBP2
Gene Aliases: 1810014B19Rik; AA537058; B830003A16Rik; C81321; EBNA1BP2; EBP2; NOBP; P40
UniProt ID: (Human) Q99848, (Mouse) Q9D903
Entrez Gene ID: (Human) 10969, (Rat) 114021, (Mouse) 69072
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support