Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody is stable for 24 months.
Positive Control: HeLa or 293T cells.Human testis, spleen or colon. Cellular Localization: Nucleus. Cytosol.
Specificity Comments: Inhibits MDM2-mediated degradation of p53/TP53 possibly by cooperating in part with TXNIP. May be involved transcriptional regulation. In vitro has intrinsic transactivation activity enhanced by EP300. May be a transcriptional activator required for the expression of glycolytic genes. Involved in regulation of cell cycle progression. Proposed to disrupt Rb-E2F binding leading to transcriptional activation of E2F proteins. The cell cycle -regulating function may depend on its RUVBL1-mediated association with the R2TP complex. May play a role in regulation of pre-mRNA splicing.
Predicted to react with Rabbit, Goat, Horse, Guinea pig, Dog, Pig, Rhesus monkey, Orangutan and Elephant (based on sequence homology)
Regulator of p53/TP53 stability and function. Inhibits MDM2-mediated degradation of p53/TP53 possibly by cooperating in part with TXNIP (PubMed:16849563, PubMed:23880345). May be involved transcriptional regulation. In vitro has intrinsic transactivation activity enhanced by EP300. May be a transcriptional activator required for the expression of glycolytic genes (PubMed:19919181, PubMed:9928932). Involved in regulation of cell cycle progression. Proposed to disrupt Rb-E2F binding leading to transcriptional activation of E2F proteins (PubMed:19640839). The cell cycle -regulating function may depend on its RUVBL1-mediated association with the R2TP complex (PubMed:26711270). May play a role in regulation of pre-mRNA splicing (PubMed:24722212). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ecdysoneless homolog; hSGT1; Human suppressor of GCR two; Protein ecdysoneless homolog; protein SGT1; SGT1 protein; suppressor of GCR2; suppressor of S. cerevisiae gcr2
Gene Aliases: ECD; GCR2; HSGT1; SGT1
UniProt ID: (Human) O95905
Entrez Gene ID: (Human) 11319
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support