Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TAAELQTLA PAINWLPFLN TIFYPVEINE SEPIVVYDKE YLEQISTLIN TTDRCLLNNY MIWNLVRKTS SFLDQRFQDA DEKFMEVMYG TK (335-425 aa encoded by BC126257)
ECE-1 is involved in proteolytic processing of endothelin precursors to biologically active peptides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ECE 1; ECE-1; Endothelin-converting enzyme 1
Gene Aliases: ECE; ECE1
UniProt ID: (Human) P42892
Entrez Gene ID: (Human) 1889
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support