Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRGVWPPPVS ALLSALGMST YKRATLDEED LVDSLSEGDA YPNGLQVNFH SPRSGQRCWA ARTQVEKRLV VLVVLLAAGL VACLAALGIQ YQTRSPSVCL SEACVSVTSS ILSSMDPTVD PCHDFFSYAC GGWIKANPVP DGHSRWGTFS NLWEHNQAII KHLLENSTAS VSEAERKAQV YYRACMNETR IEELRAKPLM ELIERLGGWN ITGPWAKDNF QDTLQVVTAH YRTSPFFSVY VSADSKNSNS NVIQVDQSGL GLPSRDYYLN KTENEKVLTG YLNYMVQLGK LLGGGDEEAI RPQMQQILDF ETALANITIP QEKRRDEELI YHKVTAAELQ TLAPAINWLP FLNTIFYPVE INESEPIVVY DKEYLEQIST LINTTDRCLL NNYMIWNLVR KTSSFLDQRF QDADEKFMEV MYGTKKTCLP RWKFCVSDTE NNLGFALGPM FVKATFAEDS KSIATEIILE IKKAFEESLS TLKWMDEETR KSAKEKADAI YNMIGYPNFI MDPKELDKVF NDYTAVPDLY FENAMRFFNF SWRVTADQLR KAPNRDQWSM TPPMVNAYYS PTKNEIVFPA GILQAPFYTR SSPKALNFGG IGVVVGHELT HAFDDQGREY DKDGNLRPWW KNSSVEAFKR QTECMVEQYS NYSVNGEPVN GRHTLGENIA DNGGLKAAYR AYQNWVKKNG AEHSLPTLGL TNNQLFFLGF AQVWCSVRTP ESSHEGLITD PHSPSRFRVI GSLSNSKEFS EHFRCPPGSP MNPPHKCEVW
ECE-1 is involved in proteolytic processing of endothelin precursors to biologically active peptides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ECE 1; ECE-1; Endothelin-converting enzyme 1
Gene Aliases: ECE; ECE1
UniProt ID: (Human) P42892
Entrez Gene ID: (Human) 1889
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support