Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In Western blots, 30% cross-reactivity with recombinant human (rh) ECE-1 and no cross-reactivity with rhKELL is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
Converts big endothelin-1 to endothelin-1. Also involved in the processing of various neuroendocrine peptides, including neurotensin, angiotensin I, substance P, proenkephalin-derived peptides, and prodynorphin-derived peptides. May limit beta-amyloid peptide accumulation in brain. May also have methyltransferase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ECE-2; EEF1AKMT4-ECE2 readthrough transcript protein; Endothelin-converting enzyme 2
Gene Aliases: ECE2; EEF1AKMT4-ECE2; KIAA0604; UNQ403/PRO740
UniProt ID: (Human) P0DPD8
Entrez Gene ID: (Human) 9718
Molecular Function:
metalloprotease
methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support