Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAGIVASR RLRDLLTRRL TGSNYPGLSI SLRLTGSSAQ EAASGVALGE APDHSYESLR VTSAQKHVLH VQLNRPNKRN AMNKVFWREM VECFNKISRD ADCRAVVISG AGKMFTAGID LMDMASDILQ PKGDDVARIS WYLRDIITRY QETFNVIERC PKPVIAAVHG GCIGGGVDLV TACDIRYCAQ DAFFQVKEVD VGLAADVGTL QRLPKVIGNQ SLVNELAFTA RKMMADEALG SGLVSRVFPD KEVMLDAALA LAAEISSKSP VAVQSTKVNL LYSRDHSVAE SLNYVASWNM SMLQTQDLVK SVQATTENKE LKTVTFSKL (1-328 aa encoded by BC017408 )
This gene encodes a member of the hydratase/isomerase superfamily. The gene product shows high sequence similarity to enoyl-coenzyme A (CoA) hydratases of several species, particularly within a conserved domain characteristic of these proteins. The encoded protein, which contains a C-terminal peroxisomal targeting sequence, localizes to the peroxisome. The rat ortholog, which localizes to the matrix of both the peroxisome and mitochondria, can isomerize 3-trans,5-cis-dienoyl-CoA to 2-trans,4-trans-dienoyl-CoA, indicating that it is a delta3,5-delta2,4-dienoyl-CoA isomerase. This enzyme functions in the auxiliary step of the fatty acid beta-oxidation pathway. Expression of the rat gene is induced by peroxisome proliferators.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Delta(3,5)-Delta(2,4)-dienoyl-CoA isomerase, mitochondrial; delta3,5-delta2,4-dienoyl-CoA isomerase; delta3,5-delta2,4-dienoyl-coenzyme A isomerase; dienoyl-CoA isomerase; enoyl CoA hydratase 1, peroxisomal; enoyl Coenzyme A hydratase 1, peroxisomal; enoyl hydratase-like protein peroxisomal; enoyl hydratase-like protein, peroxisomal; enoyl-CoA hydratase 1, peroxisomal; Peroxisomal enoyl hydratase-like protein; peroxisomal enoyl-CoA hydratase 1; peroxisomal/mitochondrial dienoyl-CoA isomerase
Gene Aliases: AA617331; ECH1; HPXEL; Pxel
UniProt ID: (Human) Q13011, (Rat) Q62651, (Mouse) O35459
Entrez Gene ID: (Human) 1891, (Rat) 64526, (Mouse) 51798
Molecular Function:
hydratase
lyase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support