Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GPLRPPVRCP AWRPFASGAN FEYIIAEKRG KNNTVGLIQL NRPKALNALC DGLIDELNQA LKIFEEDPAV GAIVLTGGDK AFAAGADIKE MQNLSFQDCY SSKFLKHWDH LTQVKKPVIA AVNGYAFGGG CELAMMCDII YAGEKAQFAQ PEILIGTIPG AGGTQRLTRA VGKSLAMEMV LTGDRISAQD AKQAGLVSKI CPVETLVEEA IQCAEKIASN SKIVVAMAKE SVNAAFEMTL TEGSKLEKKL FYSTFATDDR KEGMTAFVEK RKANFKDQ
The protein encoded by this gene functions in the second step of the mitochondrial fatty acid beta-oxidation pathway. It catalyzes the hydration of 2-trans-enoyl-coenzyme A (CoA) intermediates to L-3-hydroxyacyl-CoAs. The gene product is a member of the hydratase/isomerase superfamily. It localizes to the mitochondrial matrix. Transcript variants utilizing alternative transcription initiation sites have been described in the literature.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ECHS1; enoyl CoA hydratase, short chain, 1, mitochondrial; enoyl Coenzyme A hydratase, short chain, 1, mitochondrial; Enoyl-CoA hydratase 1; Enoyl-CoA hydratase, mitochondrial; enoyl-CoA hydratase, short chain, 1, mitochondrial; mECH; SCEH; short chain enoyl-CoA hydratase; Short-chain enoyl-CoA hydratase
Gene Aliases: ECHS1; ECHS1D; SCEH
UniProt ID: (Human) O00739
Entrez Gene ID: (Human) 1892
Molecular Function:
hydratase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support