Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: LPKENWLKML CRHVANTICK ADAENLIYTA DPESFEVNTK DMDSTLSRAS RAIKKTSKKV TRAFSFSKTP KRALRRALMT SHGSVEGRSP SSNDKHVMSR
The protein encoded by this gene is a transforming protein that is related to Rho-specific exchange factors and yeast cell cycle regulators. The expression of this gene is elevated with the onset of DNA synthesis and remains elevated during G2 and M phases. In situ hybridization analysis showed that expression is at a high level in cells undergoing mitosis in regenerating liver. Thus, this protein is expressed in a cell cycle-dependent manner during liver regeneration, and is thought to have an important role in the regulation of cytokinesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epithelial cell transforming sequence 2 oncogene; Epithelial cell-transforming sequence 2 oncogene; FLJ10461; MGC138291; Protein ECT2
Gene Aliases: ARHGEF31; ECT2
UniProt ID: (Human) Q9H8V3
Entrez Gene ID: (Human) 1894
Molecular Function:
G-protein modulator
protein-binding activity modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support