Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 2
In Western blots, less than 1% cross-reactivity with recombinant human (rh) 4-1BB, rhCD27, rhCD30, rhCD40, rhDR6, rhTAJ, rhTNF RI, rhTNF RII, rhFas, rhGITR, rhHVEM, rhLTR beta, rhNGFR, rhOPG and rhRANK is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.
This gene encodes a member of the tumor necrosis factor receptor family. The encoded transmembrane protein is a receptor for the soluble ligand ectodysplasin A, and can activate the nuclear factor-kappaB, JNK, and caspase-independent cell death pathways. It is required for the development of hair, teeth, and other ectodermal derivatives. Mutations in this gene result in autosomal dominant and recessive forms of hypohidrotic ectodermal dysplasia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anhidrotic ectodysplasin receptor 1; Downless homolog; downless, mouse, homolog of; Ectodermal dysplasia receptor; ectodysplasin 1, anhidrotic receptor; Ectodysplasin-A receptor; EDA-A1 receptor; Tumor necrosis factor receptor superfamily member EDAR
Gene Aliases: DL; ECTD10A; ECTD10B; ED1R; ED3; ED5; EDA-A1R; EDA1R; EDA3; EDAR; HRM1
UniProt ID: (Human) Q9UNE0
Entrez Gene ID: (Human) 10913
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support