Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
EDAR12 recognises and binds to CRD1 of the extracellular domain of mouse EDAR. EDAR3 cross-reacts with EDAR derived from human, dog, rat and chicken when EDAR is fused to the glycosylphosphatidylinositol anchor of TRAILR3. EDAR is the receptor for the TNF family ligand EDA1, which is a type II transmembrane protein possessing a collagen-like domain and a C-terminal TNF-homology domain. EDAR is important for the correct development of skin appendages including hair, teeth and eccrine sweat glands. LoF mutations in the Eda gene is known to cause XLHED (X-linked hypohidrotic ectodermal dysplasia), and results in abnormal development.
This gene encodes a member of the tumor necrosis factor receptor family. The encoded transmembrane protein is a receptor for the soluble ligand ectodysplasin A, and can activate the nuclear factor-kappaB, JNK, and caspase-independent cell death pathways. It is required for the development of hair, teeth, and other ectodermal derivatives. Mutations in this gene result in autosomal dominant and recessive forms of hypohidrotic ectodermal dysplasia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anhidrotic ectodysplasin receptor 1; Downless; Downless homolog; downless, mouse, homolog of; Ectodermal dysplasia receptor; ectodysplasin 1, anhidrotic receptor; ectodysplasin A1 receptor; Ectodysplasin-A receptor; EDA-A1 receptor; receptor for tumor necrosis factor superfamily member ectodysplasin A1; Tumor necrosis factor receptor superfamily member EDAR
Gene Aliases: DL; ECTD10A; ECTD10B; ED1R; ED3; ED5; EDA-A1R; EDA1R; EDA3; EDAR; HRM1; RGD1561714
UniProt ID: (Human) Q9UNE0, (Mouse) Q9R187
Entrez Gene ID: (Human) 10913, (Dog) 481305, (Chicken) 428015, (Rat) 365581, (Mouse) 13608
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support