Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: YLLFDEDNPV HKSGTRYMFT TEGHIVSVDE HLRELPWKEF FSEEGGQDQG GKSVHRPKPH ELKVINSSSN CNRVPDERRY SLPLKSIYMR QIDQMVGL
EDEM1 belongs to the glycosyl hydrolase 47 family. It extracts misfolded glycoproteins, but not glycoproteins undergoing productive folding, from the calnexin cycle. It is directly involved in endoplasmic reticulum-associated degradation (ERAD) and targets misfolded glycoproteins for degradation in an N-glycan-dependent manner.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ER degradation enhancer, mannosidase alpha-like 1; ER degradation-enhancing alpha-mannosidase-like 1; ER degradation-enhancing alpha-mannosidase-like protein 1
Gene Aliases: EDEM; EDEM1; KIAA0212
UniProt ID: (Human) Q92611
Entrez Gene ID: (Human) 9695
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support