Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene is known to suppress G-protein and mitogen-activated signal transduction in mammalian cells. The encoded protein shares significant similarity with Arabidopsis FUS6, which is a regulator of light-mediated signal transduction in plant cells. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EDF-1; Endothelial differentiation-related factor 1; MBF1; MGC9058; multiprotein bridging factor 1; Multiprotein-bridging factor 1
Gene Aliases: CFAP280; EDF-1; EDF1; MBF1
UniProt ID: (Human) O60869
Entrez Gene ID: (Human) 8721
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support