Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
EFCAB7 (EF-Hand Calcium Binding Domain 7) is a Protein Coding gene. Diseases associated with EFCAB7 include Ellis-Van Creveld Syndrome. Among its related pathways are Signaling by Hedgehog and Signaling by GPCR. Gene Ontology (GO) annotations related to this gene include calcium ion binding. An important paralog of this gene is CABP2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EF-hand calcium-binding domain-containing protein 7; RP4-534K7.1
Gene Aliases: EFCAB7; KIAA1799; mKIAA1799
UniProt ID: (Human) A8K855, (Mouse) Q8VDY4
Entrez Gene ID: (Human) 84455, (Mouse) 230500
Molecular Function:
calcium-binding protein
calmodulin-related
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support