Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PGSYQCTCP DGYRKIGPEC VDIDECRYRY CQHRCVNLPG SFRCQCEPGF QLGPNNRSCV DVNECDMGAP CEQRCFNSYG TFLCRCHQGY ELHRDGFSCS DIDECSYSSY LCQYRCVNEP GRFSCHCPQG YQLLATRLCQ DIDECESGAH QCSEAQTCVN FHGGYRCVDT NRCVEPYIQV SENRCLCPAS NPLCREQPSS IVHRYMTITS ERSVPADVFQ IQATSVYPGA YNAFQIRAGN SQGDFYIRQI NNVSAMLVLA RPVTGPREYV LDLEMVTMNS LMSYRASSVL RLTVFVGAYT F (144-443 aa encoded by BC010456)
EFEMP2 have been found to contain variations of the epidermal growth factor (EGF) domain and have been implicated in functions as diverse as blood coagulation, activation of complement and determination of cell fate during development. The protein contains four EGF2 domains and six calcium-binding EGF2 domains. This protein is necessary for elastic fiber formation and connective tissue development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EGF containing fibulin-like extracellular matrix protein 2; EGF-containing fibulin-like extracellular matrix protein 2; EGF-containing fibulin-like extracellular matrix protein 2 (Efemp2); FIBL-4; fibulin 4; Fibulin-4; Fibulin4; mutant p53 binding protein 1; Mutant p53-binding protein 1; Protein UPH1
Gene Aliases: 0610011K11Rik; ARCL1B; EFEMP2; FBLN4; MBP1; UNQ200/PRO226; UPH1
UniProt ID: (Human) O95967, (Mouse) Q9WVJ9
Entrez Gene ID: (Human) 30008, (Mouse) 58859
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support