Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SDLIRSMEQL TSSLNEGENT HMIHQKTQEK IWEFKGKLEQ HLTGRRWRNI VDQIWSFGPR KCGPNILVNK SEDFQNSVWT GPADKASKEA SRYRDLGNSI VSGFQLATL
Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.
EFTUD1 is involved in the biogenesis of the 60S ribosomal subunit and translational activation of ribosomes. Together with SBDS, triggers the GTP-dependent release of EIF6 from 60S pre-ribosomes in the cytoplasm, thereby activating ribosomes for translation competence by allowing 80S ribosome assembly and facilitating EIF6 recycling to the nucleus, where it is required for 60S rRNA processing and nuclear export. Has low intrinsic GTPase activity. GTPase activity is increased by contact with 60S ribosome subunits.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: elongation factor Tu GTP binding domain containing 1; Elongation factor Tu GTP-binding domain-containing protein 1; Elongation factor-like 1; Elongation factor-like GTPase 1; Protein FAM42A; ribosome assembly 1 homolog
Gene Aliases: EFL1; EFTUD1; FAM42A; HsT19294; RIA1
UniProt ID: (Human) Q7Z2Z2
Entrez Gene ID: (Human) 79631
Molecular Function:
translation elongation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support