Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In ELISAs, this antibody does not cross-react with recombinant mouse (rm) Amphiregulin, rmBetacellulin, rhEGF, rmEpigen, rmEpiregulin, rhHB-EGF, rhNRG-1 alpha, rhNRG1-beta 1, or rhTGF-alpha.
Reconstitute at 0.5 mg/mL in sterile PBS.
EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EGF; H-EGF; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF)
Gene Aliases: AI790464; Egf
UniProt ID: (Mouse) P01132
Entrez Gene ID: (Mouse) 13645
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support