Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: YRPGRRVCAV RAHGDPVSES FVQRVYQPFL TTCDGHRACS TYRTIYRTAY RRSPGLAPAR PRYACCPGWK RTSGLPGACG AAICQPPCRN GGSCVQPGRC RCPAGWRGDT CQSDVDERSA RRGGCPQRCI NTAGSYWCQC WEGHSLSADG TLCVPKGGPP RVAPNPTGVD SAMKEEVQRL QSRVDLLEEK LQLVLAPLHS LASQALEHGL PDPGSLLVHS FQQLGRIDSL SEQISFLEEQ LGSCSCKKDS
Epidermal growth factor-like protein 7 (EGFL7), also known as VE-statin, MEGF7, Notch4-like protein or Zneu1, is a 29.6 kDa protein produced and secreted by endothelium. The protein is also expressed by some normal and malignant tissues. The main function of EGFL7 is to regulate angiogenesis and vasculogenesis. A high level of EFGL-7 can be found in tissues with active angiogenesis of a variety of disorders.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EGF-like protein 7; EGF-like-domain, multiple 7; Epidermal growth factor-like protein 7; Multiple EGF-like domains protein 7; Multiple epidermal growth factor-like domains protein 7; NOTCH4-like protein; RP11-251M1.2; Vascular endothelial statin; VE-statin; Zneu1
Gene Aliases: EGFL7; MEGF7; NEU1; UNQ187/PRO1449; VE-STATIN; ZNEU1
UniProt ID: (Human) Q9UHF1
Entrez Gene ID: (Human) 51162
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support