Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.
Immunogen sequence: EVLGTQKTEA GGSGVLQTRT TIAETEVLVT QEISGDLGPL KIEDTIQSEM LGTQETEVEA SRVPESEAEG TEAKILGTQE ITARDSGV
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EH domain binding protein 1-like 1; EH domain-binding protein 1-like protein 1; tangerin
Gene Aliases: EHBP1L1
UniProt ID: (Human) Q8TB89
Entrez Gene ID: (Human) 254102
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support