Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: KRMQDQLQAQ DFSKFQPLKS KLLEVVDDML AHDIAQLMVL VRQEESQRPI
EHD3 plays a role in endocytic transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EH domain-containing protein 3; EH-domain containing 3; PAST homolog 3
Gene Aliases: EHD2; EHD3; PAST3
UniProt ID: (Human) Q9NZN3
Entrez Gene ID: (Human) 30845
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support