Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MFSWMGRQAG GRERAGGADA VQTVTGGLRS LYLRKVLPLE EAYRFHEFHS PALEDADFEN KPMILLVGQY STGKTTFIRY LLEQDFPGMR IGPEPTTDSF IAVMYGETEG STPGNALVVD PKKPFRKLSR FGNAFLNRFM CSQLPNQVLK SISVIDSPGI LSGEKQRISR GYDFCQVLQW FAERVDRIIL LFDAHKLDIS DEFSEAIKAF RGQDDKIRVV LNKADQVDTQ QLMRVYGALM WSLGKVINTP EVLRVYIGSF WAQPLQNTDN RRLFEAEAQD LFRDIQSLPQ KAAVRKLNDL IKRARLAKVH AYIISYLKKE MPSVFGKENK KRELISRLPE IYIQLQREYQ ISAGDFPEVK AMQEQLENYD FTKFHSLKPK LIEAVDNMLS NKISPLMNLI SQEETSTPTQ LVQGGAFDGT TEGPFNQGYG EGAKEGADEE EWVVAKDKPV YDELFYTLSP INGKISGVNA KKEMVTSKLP NSVLGKIWKL ADCDCDGMLD EEEFALAKHL IKIKLDGYEL PSSLPPHLVP PSHRKSLPKA D
ATP- and membrane-binding protein that probably controls membrane reorganization/tubulation upon ATP hydrolysis. Plays a role in early endosomal transport. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EH domain-containing protein 4; EH-domain containing 4; Hepatocellular carcinoma-associated protein 10/11; hepatocellular carcinoma-associated protein HCA11; ortholog of rat pincher; PAST homolog 4
Gene Aliases: EHD4; FKSG7; HCA10; HCA11; PAST4
UniProt ID: (Human) Q9H223
Entrez Gene ID: (Human) 30844
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support