Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MILEGGGVMN LNPGNNLLHQ PPAWTDSYST CNVSSGFFGG QWHEIHPQYW TKYQVWEWLQ HLLDTNQLDA NCIPFQEFDI NGEHLCSMSL QEFTRAAGTA GQLLYSNLQH LKWNGQCSSD LFQSTHNVIV KTEQTEPSIM NTWKDENYLY DTNYGSTVDL LDSKTFCRAQ ISMTTTSHLP VAESPDMKKE QDPPAKCHTK KHNPRGTHLW EFIRDILLNP DKNPGLIKWE DRSEGVFRFL KSEAVAQLWG KKKNNSSMTY EKLSRAMRYY YKREILERVD GRRLVYKFGK NARGWRENEN
This gene encodes a protein that belongs to an ETS transcription factor subfamily characterized by epithelial-specific expression (ESEs). The encoded protein acts as a transcriptional repressor and may be associated with asthma susceptibility. This protein may be involved in epithelial differentiation and carcinogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EHF; Epithelium-specific Ets transcription factor 3; ESE-3; ESE3 transcription factor; ETS domain-containing transcription factor; ETS homologous factor; hEHF
Gene Aliases: EHF; ESE3; ESE3B; ESEJ
UniProt ID: (Human) Q9NZC4
Entrez Gene ID: (Human) 26298
Molecular Function:
winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support