Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MKMDVSVRAA GCSDDLSSGE ADVDPKLLEL TADEEKCRSI RRQYRQLMYC VRQNREDIVS SANNSLTEAL EEANVLFDGV SRTREAALDA RFLVMASDLG KEKAKQLNSD MNFFNQLAFC DFLFLFVGLN WMEGDPDKLS DCDDSIALSF WKAIEKEATS WMVKAETFHF VFGSFKLERS APKPRLEHQK KVRKMEENGN MPTKLQKLDL SSYPEATEKN VERILGLLQT YFRKYPDTPV SYFEFVIDPN SFSRTVENIF YVSFIVRDGF ARIRLDEDRL PILEPMNVNQ MGEGNDSSCH GRKQGVISLT LQEWKNIVAA FEISEAMITY SSY
Tissue-specific component of the SMC5-SMC6 complex, a complex involved in repair of DNA double-strand breaks by homologous recombination. The complex may promote sister chromatid homologous recombination by recruiting the SMC1-SMC3 cohesin complex to double-strand breaks. The complex is required for telomere maintenance via recombination and mediates sumoylation of shelterin complex (telosome) components. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E1A-like inhibitor of differentiation 3; EID-1-like inhibitor of differentiation 3; EID-3; EID3; EP300-interacting inhibitor of differentiation 3; non-SMC element 4 homolog B; Non-structural maintenance of chromosomes element 4 homolog B; NS4EB; testis tissue sperm-binding protein Li 96mP
Gene Aliases: EID3; NS4EB; NSE4B; NSMCE4B
UniProt ID: (Human) Q8N140
Entrez Gene ID: (Human) 493861
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support