Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Necessary for scanning and involved in initiation site selection. Promotes the assembly of 48S ribosomal complexes at the authentic initiation codon of a conventional capped mRNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A121; eIF1; eIF13; Eukaryotic translation initiation factor 1; Protein translation factor SUI1 homolog; Sui1iso1
Gene Aliases: A121; EIF-1; EIF1; EIF1A; ISO1; SUI1
UniProt ID: (Human) P41567
Entrez Gene ID: (Human) 10209
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support