Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31725 has been tested in indirect ELISA.
EIF2A is a 65-kD protein that catalyzes the formation of puromycin-sensitive 80S preinitiation complexes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 65 kDa eukaryotic translation initiation factor 2A; eIF-2A; Eukaryotic translation initiation factor 2A; eukaryotic translation initiation factor 2A, 65kDa
Gene Aliases: CDA02; EIF-2A; EIF2A; MST089; MSTP004; MSTP089
UniProt ID: (Human) Q9BY44
Entrez Gene ID: (Human) 83939
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support