Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: IASTDVDKTG ASYYGEQTLH YIATNGESAV VQLPKNGPIY DVVWNSSSTE FCAVYGFMPA KATIFNLKCD PVFDFGTGPR NAAYYSPHGH ILVLAGFGNL RGQMEVWDVK NYKLISKPVA SDSTYFAWCP DGEHILTATC APRLRVNNGY KIWHYTGSIL HKYDVPSNAE LWQVSWQPFL DGIFPAKTIT YQAVPSEVPN EEPKVATAYR PPALRNKPIT NSKLHEEEPP QNMKPQSGND KPLSKTALKN QRKHEAKKAA KQEARSDKSP DLAPTPAPQS TPRNTVSQSI SGDPEIDKKI KNLKKKLKAI EQLKEQAATG KQLEKNQLEK IQKETALLQE LEDLKLGI
EIF2A is a 65-kD protein that catalyzes the formation of puromycin-sensitive 80S preinitiation complexes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 65 kDa eukaryotic translation initiation factor 2A; eIF-2A; Eukaryotic translation initiation factor 2A; eukaryotic translation initiation factor 2A, 65kDa
Gene Aliases: CDA02; D030048D22; D3Ertd194e; EIF-2A; EIF2A; MST089; MSTP004; MSTP089
UniProt ID: (Human) Q9BY44, (Mouse) Q8BJW6
Entrez Gene ID: (Human) 83939, (Rat) 502531, (Mouse) 229317
Molecular Function:
translation factor
translation initiation factor
translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support