Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FAEPTPEKEK RFGESDTENQ NNKSVKYTTN LVIRESGELE STLELQENGL AGLSASSIVE QQLPLRRNSH LEESFTSTEE SSEENVNFLG QTEAQYHLML HIQMQLCELS LWDWIVERNK RGREYVDESA CPYVMANVAT KIFQELVEGV FYIHNMGIVH RDLKPRNIFL HGPDQQVKIG DFGLACTDIL QKNTDWTNRN GKRTPTHTSR VGTCLYASPE QLEGSEYDAK SDMYSLGVVL LELFQPFGTE MERAEVLTGL RTGQLPESLR KRCPVQAKYI QHLTRRNSSQ RPSAIQLLQS ELFQNSGNVN LTLQMKIIEQ EKEIAELKKQ LNLLSQDKGV RDDGKDGGVG
EIF2AK1 (HRI) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK1, signaling is initiated by low levels of heme. The HRI gene is localized to 7p22 where its 3' end slightly overlaps the 3' end of the gene JTV1. The two genes are transcribed from opposite strands and kinase activity is induced by low levels of heme and inhibited by the presence of heme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Eukaryotic translation initiation factor 2-alpha kinase 1; HCR; he; heme regulated initiation factor 2 alpha kinase; heme sensitive initiation factor 2a kinase; Heme-controlled repressor; Heme-regulated eukaryotic initiation factor eIF-2-alpha kinase; Heme-regulated inhibitor; heme-regulated initiation factor 2-alpha kinase; heme-regulated repressor; Hemin-sensitive initiation factor 2-alpha kinase; hHRI
Gene Aliases: EIF2AK1; HCR; HRI; KIAA1369; PRO1362
UniProt ID: (Human) Q9BQI3
Entrez Gene ID: (Human) 27102
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support