Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPGSAAKGS ELSERIESFV ETLKRGGGPR SSEEMARETL GLLRQIITDH RWSNAGELME LIRREGRRMT AAQPSETTVG NMVRRVLKII REEYGRLHGR SDESDQQESL HKLLTSGGLN EDFSFHYAQL QSNIIEAINE LLVELEGTME NIAAQALEHI HSNEVIMTIG FSRTVEAFLK EAARKRKFHV IVAECAPFCQ GHEMAVNLSK AGIETTVMTD AAIFAVMSRV NKVIIGTKTI LANGALRAVT GTHTLALAAK HHSTPLIVCA PMFKLSPQFP NEEDSFHKFV APEEVLPFTE GDILEKVSVH CPVFDYVPPE LITLFISNIG GNAPSYIYRL MSELYHPDDH VL (1-351 aa encoded by BC011750)
eIF2b beta is the beta subunit of eukaryotic initiation factor-2B (EIF2B). EIF2B is involved in protein synthesis and exchanges GDP and GTP for its activation and deactivation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-2B GDP-GTP exchange factor subunit beta; eIF2B GDP-GTP exchange factor subunit beta; eukaryotic translation initiation factor 2B, subunit 2 (beta, 39kD); eukaryotic translation initiation factor 2B, subunit 2 beta, 39kDa; S20I15; S20III15; translation initiation factor eIF-2B subunit beta; Translation initiation factor eIF2B subunit beta
Gene Aliases: AA409345; C85417; EIF-2Bbeta; EIF2B; EIF2B2; EIF2BB
UniProt ID: (Human) P49770, (Mouse) Q99LD9
Entrez Gene ID: (Human) 8892, (Mouse) 217715, (Rat) 84005
Molecular Function:
translation initiation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support