Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
AGO1 is a member of the Argonaute family of proteins which play a role in RNA interference. The protein is highly basic, and contains a PAZ domain and a PIWI domain. It may interact with dicer1 and play a role in short-interfering-RNA-mediated gene silencing. AGO1 binds to miRNAs or siRNAs and represses the translation of mRNAs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: argonaute 1; Argonaute RISC catalytic component 1; Argonaute1; eIF-2C 1; eIF2C 1; Eukaryotic translation initiation factor 2C 1; eukaryotic translation initiation factor 2C, 1; Golgi Endoplasmic Reticulum protein 95 kDa; Protein argonaute-1; Putative RNA-binding protein Q99
Gene Aliases: AGO1; EIF2C; EIF2C1; GERP95; hAgo1; Q99
UniProt ID: (Human) Q9UL18
Entrez Gene ID: (Human) 26523
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support