Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SVAGEYGRHS LYKMLGYFSL VGLLRLHSLL GDYYQAIKVL ENIELNKKSM YSRVPECQVT TYYYVGFAYL MMRRYQDAIR VFANILLYIQ RTKSMFQRTT YKYEMINKQN EQMHALLAIA LTMYPMRIDE SIHLQLREKY GDKMLRMQKG DPQVYEELFS YSCPKFLSPV VPNYDNVHPN YHKEPFLQQL KVFSDEVQQQ AQLSTIRSFL KLYTTMPVAK LAGFLDLTEQ EFRIQLLVFK HKMKNLVWTS GISALDGEFQ SASEVDFYID KDMIHIADTK VARRYGDFFI RQIHKFEELN RTLKKMGQRP
Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis (PubMed:17581632, PubMed:25849773, PubMed:27462815). The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S pre-initiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post-termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation (PubMed:17581632). The eIF-3 complex specifically targets and initiates translation of a subset of mRNAs involved in cell proliferation, including cell cycling, differentiation and apoptosis, and uses different modes of RNA stem-loop binding to exert either translational activation or repression (PubMed:25849773). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 66 kDa tyrosine-rich heat shock protein; 67 kDa polymerase-associated factor; eIEF associated protein HSPC021; EIF3L; Eukaryotic translation initiation factor 3 subunit 6-interacting protein; Eukaryotic translation initiation factor 3 subunit E-interacting protein; Eukaryotic translation initiation factor 3 subunit L; eukaryotic translation initiation factor 3, subunit L; HSP-66Y; PAF67
Gene Aliases: 0610011H21Rik; D15N1e; EIF3EIP; Eif3ip; EIF3L; EIF3S11; EIF3S6IP; HSP-66Y; HSPC021; HSPC025; MSTP005; PAF67
UniProt ID: (Human) Q9Y262, (Mouse) Q8QZY1
Entrez Gene ID: (Human) 51386, (Mouse) 223691
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support