Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNNKFDALK DDDSGDHDQN EENSTQKDGE KEKTERDKNQ SSSKRKAVVP GPAEHPLQYN YTFWYSRRTP GRPTSSQSYE QNIKQIGTFA SVEQFWRFYS HMVRPGDLTG HSDFHLFKEG IKPMWEDDAN KNGGKWIIRL RKGLASRCWE NLILAMLGEQ FMVGEEICGA VVSVRFQEDI ISIWNKTASD QATTARIRDT LRRVLNLPPN TIMEYKTHTD SIKMPGRLGP QRLLFQNLWK PRLNVP (1-245 aa encoded by B C021690)
Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation (PubMed:9582349, PubMed:17368478, PubMed:25624349). Acts as a repressor of translation initiation (PubMed:22751931). In contrast to EIF4E, it is unable to bind eIF4G (EIF4G1, EIF4G2 or EIF4G3), suggesting that it acts by competing with EIF4E and block assembly of eIF4F at the cap. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-4E type 2; eIF4E type 2; eIF4E-2; eIF4E-like cap-binding protein; eIF4E-like protein 4E-LP; eukar; Eukaryotic translation initiation factor 4E homologous protein; eukaryotic translation initiation factor 4E like 3; eukaryotic translation initiation factor 4E member 2; Eukaryotic translation initiation factor 4E type 2; Eukaryotic translation initiation factor 4E-like 3; h4EHP; mRNA cap-binding protein 4EHP; mRNA cap-binding protein type 2; mRNA cap-binding protein type 3
Gene Aliases: 2700069E09Rik; 4E-LP; 4EHP; AI036339; AV129531; D0H0S6743E; EIF4E2; EIF4EL3; IF4e
UniProt ID: (Human) O60573, (Mouse) Q8BMB3
Entrez Gene ID: (Human) 9470, (Rat) 363275, (Mouse) 26987
Molecular Function:
translation factor
translation initiation factor
translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support