Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MNNKFDALKD DDSGDHDQNE ENSTQKDGEK EKTERDKNQS SSKRKAVVPG PAEHPLQYNY TFWYSRRTPG RPTSSQSYEQ NIKQIGTFAS VEQFWRFYSH MVRPGDLTGH SDFHLFKEGI KPMWEDDANK NGGKWIIRLR KGLASRCWEN LILAMLGEQF MVGEEICGAV VSVRFQEDII SIWNKTASDQ ATTARIRDTL RRVLNLPPNT IMEYKTHTDS IKMPGRLGPQ RLLFQNLWKP RLNVP
Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation (PubMed:9582349, PubMed:17368478, PubMed:25624349). Acts as a repressor of translation initiation (PubMed:22751931). In contrast to EIF4E, it is unable to bind eIF4G (EIF4G1, EIF4G2 or EIF4G3), suggesting that it acts by competing with EIF4E and block assembly of eIF4F at the cap. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-4E type 2; eIF4E-like cap-binding protein; eIF4E-like protein 4E-LP; eukar; Eukaryotic translation initiation factor 4E homologous protein; Eukaryotic translation initiation factor 4E type 2; Eukaryotic translation initiation factor 4E-like 3; h4EHP; mRNA cap-binding protein 4EHP; mRNA cap-binding protein type 3
Gene Aliases: 4E-LP; 4EHP; EIF4E2; EIF4EL3; IF4e
UniProt ID: (Human) O60573
Entrez Gene ID: (Human) 9470
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support