Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
EIF4E3 belongs to the EIF4E family of translational initiation factors that interact with the 5-prime cap structure of mRNA and recruit mRNA to the ribosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-4E type 3; Eukaryotic translation initiation factor 4E type 3
Gene Aliases: eIF-4E3; eIF4E-3; EIF4E3
UniProt ID: (Human) Q8N5X7
Entrez Gene ID: (Human) 317649
Molecular Function:
translation factor
translation initiation factor
translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support