Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MSTSTSCPIP GGRDQLPDCY STTPGGTLYA TTPGGTRIIY DRKFLLECKN SPIARTPPCC LPQIPGVTTP PTAPLSKLEE LKEQETEEEI PDDAQFEMDI
This gene encodes a member of the EIF4EBP family which derives it name from proteins that bind to eukaryotic initiation factor 4E and that prevent its assembly into EIF4F. Co-transcription of this gene and the neighboring upstream gene (MASK) generates a transcript (MASK-BP3) which encodes a fusion protein comprised of the MASK protein sequence for the majority of the protein and a different C-terminus due to an alternate reading frame for the EIF4EBP3 segments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4E-BP3; eIF4E-binding protein 3; eukaryotic initiation factor 4E-binding protein 3; Eukaryotic translation initiation factor 4E-binding protein 3; NG36
Gene Aliases: 4E-BP3; 4EBP3; EIF4EBP3
UniProt ID: (Human) O60516
Entrez Gene ID: (Human) 8637
Molecular Function: translation initiation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support