Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPPPADIVKV AIEWPGAYPK LMEIDQKKPL SAIIKEVCDG WSLANHEYFA LQHADSSNFY ITEKNRNEIK NGTILRLTTS PAQNAQQLHE RIQSSSMDAK LEALKDLASL SRDVTFAQEF INLDGISLLT QMVESGTERY QKLQKIMKPC FGDMLSFTLT AFVELMDHGI VSWDTFSVAF IKKIASFVNK SAIDISILQR SLAILESMVL NSHDLYQKVA QEITIGQLIP HLQGSDQEIQ TYTIAVINAL FLKAPDERRQ EMANILAQKQ LRSIILTHVI RAQRAINNEM AHQLYVLQVL TFNLLEDRMM TKMDPQDQAQ RDIIFELRRI AFDAESEPNN SSGSMEKRKS MYTRDYKKLG FINHVNPAMD FTQTPPGMLA LDNMLYFAKH HQDAYIRIVL ENSSREDKHE CPFGRSSIEL TKMLCEILKV GELPSETCND FHPMFFTHDR SFEEFFCICI QLLNKTWKEM RATSEDFNKV MQVVKEQVMR ALTTKPSSLD QFKSKLQNLS YTEILKIRQS ERMNQEDFQS RPILELKEKI QPEILELIKQ QRLNRLVEGT CFRKLNARRR QDKFWYCRLS PNHKVLHYGD LEESPQGEVP HDSLQDKLPV ADIKAVVTGK DCPHMKEKGA LKQNKEVLEL AFSILYDSNC QLNFIAPDKH EYCIWTDGLN ALLGKDMMSD LTRNDLDTLL SMEIKLRLLD LENIQIPDAP PPIPKEPSNY DFVYDCN
The protein encoded by this gene interacts with the dedicator of cyto-kinesis 1 protein to promote phagocytosis and effect cell shape changes. Similarity to a C. elegans protein suggests that this protein may function in apoptosis and in cell migration. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ced-12 homolog 1; Engulfment and cell motility protein 1; MGC126406; OTTHUMP00000207056; OTTHUMP00000207060; Protein ced-12 homolog
Gene Aliases: CED-12; CED12; ELMO-1; ELMO1; KIAA0281
UniProt ID: (Human) Q92556
Entrez Gene ID: (Human) 9844
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support