Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MADLGATKDR IISEINRLQQ EATEHATESE GRFRGLEEGQ AQAGQCPSLE GRLGRLEGVC ERLDTVAGGL QGLREGLSRH VAGLWAGLRE TNTTSQMQAA LLEKLVGGQA GLGRRLGALN SSLQLLEDRL HQLSLKDLTG EGTKGPAGEA GPPGPPGLQG PPGPAGPPGS PGKDGQEGPI GPPGPQGEQG VEGAPAAPVP QVAFSAALSL PRSEPGTVPF DRVLLNDGGY YDPETGVFTA PLAGRYLLSA VLTGHRHEKV EAVLSRSNQG VARVDSGGYE PEGLEKKPVA ESQPSPGTLG VFSLILPLQA GDTVCVDLVM GQLAHSEEPL TIFSGALLYG DPELEHA
This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal dominant cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Elastin microfibril interface-located protein 1; Elastin microfibril interfacer 1; EMILIN-1
Gene Aliases: EMI; EMILIN; EMILIN1; gp115
UniProt ID: (Human) Q9Y6C2
Entrez Gene ID: (Human) 11117
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support